2-Fluor-5-methylphenylisocyanat

Lieferant: Thermo Scientific

Synonyme: 1-Fluor-2-isocyanato-4-methylbenzol, 4-Fluor-3-isocyanatotoluol

AC31931.1
MAYBAC31931.1EA 0 EUR
MAYBAC31931.1
2-Fluor-5-methylphenylisocyanat
2-Fluor-5-methylphenylisocyanat
Formel: C₈H₆FNO
Molecular Weight: 151,14 g/mol
Siedepunkt: 80 °C (3 mmHg)
Lagertemperatur: Gefrierschrank
MDL: MFCD00673050
CAS-Nummer: 190774-50-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR