4-(Methylthio)phenylisothiocyanat

Lieferant: Thermo Scientific

AC31953.250
MAYBAC31953.250EA 0 EUR
MAYBAC31953.250
4-(Methylthio)phenylisothiocyanat
4-(Methylthio)phenylisothiocyanat
Formel: C₈H₇NS₂
Molecular Weight: 181,28 g/mol
Siedepunkt: 127…128 °C (1 mmHg)
Schmelzpunkt: 60…62 °C
MDL: MFCD00041368
CAS-Nummer: 15863-41-9
UN: 2923
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR