2-Methyl-3-furancarbonsäure

Lieferant: Thermo Scientific

Synonyme: 2-Methylfuran-3-carboxylic acid

AC33015.10
MAYBAC33015.10EA 0 EUR
MAYBAC33015.10
2-Methyl-3-furancarbonsäure
2-Methyl-3-furoic acid
Formel: C₆H₆O₃
Molecular Weight: 126,11 g/mol
MDL: MFCD00092314
CAS-Nummer: 6947-94-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR