2-Chlor-5-methylpyridin 94%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Chloro-5-picoline
AC34403.50
MAYBAC34403.50EA
0
EUR
MAYBAC34403.50
2-Chlor-5-methylpyridin 94%
2-Chlor-5-methylpyridin
Formel:
C₆H₆ClN Molecular Weight: 127,57 g/mol Siedepunkt: 97 °C (30 mmHg) Dichte: 1,169 g/cm³ Flammpunkt: 90 °C |
MDL:
MFCD00792460 CAS-Nummer: 18368-64-4 EINECS: 418-050-0 |
Spezifikation Testergebnisse
Appearance | Clear light yellow to straw yellow to pink-orange Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5260 to 1.5300 (20°C, 589 nm) |
GC | ≥92 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...