2-Brom-3-methylpyridin 95%

Lieferant: Thermo Scientific
Achtung

AC35766.1 AC35766.10 AC35766.50 357660050
MAYBAC35766.1EA 0 EUR
MAYBAC35766.1 MAYBAC35766.10 MAYBAC35766.50 ACRO357660050
2-Brom-3-methylpyridin 95%
2-Brom-3-methylpyridin
Formel: C₆H₆BrN
Flammpunkt: >110 °C
MDL: MFCD00239380
CAS-Nummer: 3430-17-9

Bestellen

Spezifikation Testergebnisse

Appearance Clear light yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5650 to 1.5700 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR