3-Bromphenylboronsäure 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (3-Bromphenyl)metaborsäure, Dihydroxy(3-bromphenyl)boran, 3-Brombenzolboronsäure

AC35894.1 358940010 358940050
MAYBAC35894.1EA 0 EUR
MAYBAC35894.1 ACRO358940010 ACRO358940050
3-Bromphenylboronsäure 97%
3-Bromphenylboronsäure
Formel: C₆H₆BBrO₂
Molecular Weight: 200.83 g/mol
Schmelzpunkt: 175…177 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00239386
CAS-Nummer: 89598-96-9

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR