2,4-Difluorphenylboronsäure 98%

Lieferant: Thermo Scientific
Achtung

AC35913.1 AC35913.10 AC35913.50 359130010 359130100 359130500
MAYBAC35913.1EA 0 EUR
MAYBAC35913.1 MAYBAC35913.10 MAYBAC35913.50 ACRO359130010 ACRO359130100 ACRO359130500
2,4-Difluorphenylboronsäure 98%
2,4-Difluorphenylboronsäure
Formel: C₆H₅BF₂O₂
Molecular Weight: 157.91 g/mol
Schmelzpunkt: 247…250 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01318998
CAS-Nummer: 144025-03-6

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to light beige Powder or crystals
Infrared spectrum Conforms
Melting point 247 to 251 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR