3-(Ethoxycarbonyl)phenylboronsäure

Lieferant: Thermo Scientific

Synonyme: 3-(Ethoxycarbonyl)benzeneboronic acid, 3-Ethoxycarbonylphenylboronic acid

AC35930.250 AC35930.1
MAYBAC35930.250EA 0 EUR
MAYBAC35930.250 MAYBAC35930.1
3-(Ethoxycarbonyl)phenylboronsäure
3-(Ethoxycarbonyl)phenylboronsäure
Formel: C₉H₁₁BO₄
Molecular Weight: 193.99 g/mol
MDL: MFCD02179444
CAS-Nummer: 4334-87-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR