2-Formylphenylboronsäure 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Boronobenzaldehyd, (2-Formylphenyl)metaborsäure, Dihydroxy(2-formylphenyl)boran, 2-Formylbenzolboronsäure

AC35936.1 AC35936.10 359360010 359360050
MAYBAC35936.1EA 0 EUR
MAYBAC35936.1 MAYBAC35936.10 ACRO359360010 ACRO359360050
2-Formylphenylboronsäure 98%
2-Formylphenylboronsäure
Formel: C₇H₇BO₃
Molecular Weight: 149.94 g/mol
Schmelzpunkt: 120…122 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00151822
CAS-Nummer: 40138-16-7

Bestellen

Spezifikation Testergebnisse

Appearance White to pink to orange Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR