2-Formyl-5-methoxyphenylboronsäure

Lieferant: Thermo Scientific

Synonyme: 5-Methoxy-2-formylphenylboronic acid

AC35953.250 AC35953.1
MAYBAC35953.250EA 0 EUR
MAYBAC35953.250 MAYBAC35953.1
2-Formyl-5-methoxyphenylboronsäure
2-Formyl-5-methoxyphenylboronsäure
Formel: C₈H₉BO₄
Molecular Weight: 179.9657 g/mol
MDL: MFCD03001335
CAS-Nummer: 40138-18-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR