2-Formyl-5-methoxyphenylboronsäure
Lieferant: Thermo Scientific
Synonyme:
5-Methoxy-2-formylphenylboronic acid
AC35953.250
AC35953.1
MAYBAC35953.250EA
0
EUR
MAYBAC35953.250
MAYBAC35953.1
2-Formyl-5-methoxyphenylboronsäure
2-Formyl-5-methoxyphenylboronsäure
Formel:
C₈H₉BO₄ Molecular Weight: 179.9657 g/mol |
MDL:
MFCD03001335 CAS-Nummer: 40138-18-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...