Benzothiazol-2-ol

Lieferant: Thermo Scientific

AC40197DA AC40197EA AC40197.50
MAYBAC40197DAEA 0 EUR
MAYBAC40197DA MAYBAC40197EA MAYBAC40197.50
Benzothiazol-2-ol
Benzothiazol-2-ol
Formel: C₇H₅NOS
MDL: MFCD00022868
CAS-Nummer: 934-34-9
EINECS: 213-281-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR