1-(4-Fluorbenzyl)homopiperazin
Lieferant: Thermo Scientific
AW00512.0001
MAYBAW00512.0001EA
0
EUR
MAYBAW00512.0001
1-(4-Fluorbenzyl)homopiperazin
1-(4-Fluorbenzyl)homopiperazin
Formel:
C₁₂H₁₇FN₂ Molecular Weight: 208,28 g/mol |
MDL:
MFCD00816588 CAS-Nummer: 76141-89-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...