4-[(2-Chlor-6-fluorbenzyl)oxy]-3-methoxybenzaldehyd

Lieferant: Thermo Scientific

AW01128DA AW01128.10
MAYBAW01128DAEA 0 EUR
MAYBAW01128DA MAYBAW01128.10
4-[(2-Chlor-6-fluorbenzyl)oxy]-3-methoxybenzaldehyd
4-[(2-Chlor-6-fluorbenzyl)oxy]-3-methoxybenzaldehyd
Formel: C₁₅H₁₂ClFO₃
MDL: MFCD01590672
CAS-Nummer: 306934-75-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR