Methyl-3-chlor-2-benzo[b]thiophencarboxylat

Lieferant: Thermo Scientific

BTB00312.1 BTB00312.10
MAYBBTB00312.1EA 0 EUR
MAYBBTB00312.1 MAYBBTB00312.10
Methyl-3-chlor-2-benzo[b]thiophencarboxylat
Methyl-3-chlor-2-benzo[b]thiophencarboxylat
Formel: C₁₀H₇ClO₂S
MDL: MFCD00052503
CAS-Nummer: 21211-07-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR