2-Chlorbenzhydrazid

Lieferant: Thermo Scientific

Synonyme: 2-Chlorobenzoic hydrazide

BTB00503-0.25
MAYBBTB00503-0.25EA 0 EUR
MAYBBTB00503-0.25
2-Chlorbenzhydrazid
2-Chlorbenzhydrazid
Formel: C₇H₇ClN₂O
Molecular Weight: 170,6 g/mol
MDL: MFCD00007597
CAS-Nummer: 5814-05-1
EINECS: 227-380-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR