4-Morpholinoanilin

Lieferant: Thermo Scientific

Synonyme: 4-(4-Morpholinyl)aniline

BTB01373.1 BTB01373EA BTB01373.50
MAYBBTB01373.1EA 0 EUR
MAYBBTB01373.1 MAYBBTB01373EA MAYBBTB01373.50
4-Morpholinoanilin
4-Morpholinoanilin
Formel: C₁₀H₁₄N₂O
Molecular Weight: 178.23 g/mol
MDL: MFCD00006169
CAS-Nummer: 2524-67-6
EINECS: 219-760-1
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR