4-Morpholinoanilin
Lieferant: Thermo Scientific
Synonyme:
4-(4-Morpholinyl)aniline
BTB01373.1
BTB01373EA
BTB01373.50
MAYBBTB01373.1EA
0
EUR
MAYBBTB01373.1
MAYBBTB01373EA
MAYBBTB01373.50
4-Morpholinoanilin
4-Morpholinoanilin
Formel:
C₁₀H₁₄N₂O Molecular Weight: 178.23 g/mol |
MDL:
MFCD00006169 CAS-Nummer: 2524-67-6 EINECS: 219-760-1 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...