5-Fluorisatin

Lieferant: Thermo Scientific
Achtung

BTB01627.1 BTB01627.50
MAYBBTB01627.1EA 0 EUR
MAYBBTB01627.1 MAYBBTB01627.50
5-Fluorisatin
5-Fluorisatin
Formel: C₈H₄FNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00022795
CAS-Nummer: 443-69-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR