N-[2-Nitro-4-(trifluormethyl)phenyl]piperidin

Lieferant: Thermo Scientific

Synonyme: 1-(2-Nitro-4-(trifluoromethyl)phenyl)piperidine

BTB01842.1
MAYBBTB01842.1EA 0 EUR
MAYBBTB01842.1
N-[2-Nitro-4-(trifluormethyl)phenyl]piperidin
N-[2-Nitro-4-(trifluormethyl)phenyl]piperidin
Formel: C₁₂H₁₃F₃N₂O₂
Molecular Weight: 274,240 g/mol
MDL: MFCD00052908
CAS-Nummer: 1692-79-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR