N-[2-Nitro-4-(trifluormethyl)phenyl]morpholin

Lieferant: Thermo Scientific

BTB01843.50
MAYBBTB01843.50EA 0 EUR
MAYBBTB01843.50
N-[2-Nitro-4-(trifluormethyl)phenyl]morpholin
N-[2-Nitro-4-(trifluormethyl)phenyl]morpholin
Formel: C₁₁H₁₁F₃N₂O₃
MDL: MFCD00052375
CAS-Nummer: 62054-72-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR