2-(1-Piperidinyl)anilin

Lieferant: Thermo Scientific

Synonyme: 2-(1-Piperidino)aniline

BTB02217DA BTB02217EA BTB02217.50
MAYBBTB02217DAEA 0 EUR
MAYBBTB02217DA MAYBBTB02217EA MAYBBTB02217.50
2-(1-Piperidinyl)anilin
2-(1-Piperidinyl)anilin
Formel: C₁₁H₁₆N₂
MDL: MFCD00047467
CAS-Nummer: 39643-31-7
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR