2-(1-Piperidinyl)anilin
Lieferant: Thermo Scientific
Synonyme:
2-(1-Piperidino)aniline
BTB02217DA
BTB02217EA
BTB02217.50
MAYBBTB02217DAEA
0
EUR
MAYBBTB02217DA
MAYBBTB02217EA
MAYBBTB02217.50
2-(1-Piperidinyl)anilin
2-(1-Piperidinyl)anilin
Formel:
C₁₁H₁₆N₂ |
MDL:
MFCD00047467 CAS-Nummer: 39643-31-7 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...