4-Brom-2,6-dichloranilin

Lieferant: Thermo Scientific
Achtung

BTB03106.1 BTB03106.10 BTB03106.50
MAYBBTB03106.1EA 0 EUR
MAYBBTB03106.1 MAYBBTB03106.10 MAYBBTB03106.50
4-Brom-2,6-dichloranilin
4-Brom-2,6-dichloranilin
Formel: C₆H₄BrCl₂N
Molecular Weight: 240.91 g/mol
Siedepunkt: 290
Schmelzpunkt: 90
MDL: MFCD00025162
CAS-Nummer: 697-88-1
EINECS: 211-809-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR