5-Amino-2-(1-pyrrolyl)benzonitril

Lieferant: Thermo Scientific

BTB03126.0001
MAYBBTB03126.0001EA 0 EUR
MAYBBTB03126.0001
5-Amino-2-(1-pyrrolyl)benzonitril
5-Amino-2-(1-pyrrolyl)benzonitril
Formel: C₁₁H₉N₃
CAS-Nummer: 106981-51-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR