2-Amino-5-brompyridin
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Brompyridin-2-amin, 5-Brom-2-pyridylamin
BTB03304.1
BTB03304.10
BTB03304.50
MAYBBTB03304.1EA
0
EUR
MAYBBTB03304.1
MAYBBTB03304.10
MAYBBTB03304.50
2-Amino-5-brompyridin
2-Amino-5-brompyridin
Formel:
C₅H₅BrN₂ Molecular Weight: 173.01 g/mol Siedepunkt: 244 Schmelzpunkt: 136…139 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006323 CAS-Nummer: 1072-97-5 EINECS: 214-019-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...