5-Chlor-4-fluor-2-nitroanilin

Lieferant: Thermo Scientific

Synonyme: 1-Amino-3-chlor-4-fluor-6-nitrobenzene

BTB03668.10
MAYBBTB03668.10EA 0 EUR
MAYBBTB03668.10
5-Chlor-4-fluor-2-nitroanilin
5-Chlor-4-fluor-2-nitroanilin
Formel: C₆H₄ClFN₂O₂
Molecular Weight: 190,56 g/mol
Schmelzpunkt: 146…147 °C
MDL: MFCD00052698
CAS-Nummer: 104222-34-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR