3-Chlor-4-fluorbenzoesäure

Lieferant: Thermo Scientific

BTB03715.1 BTB03715.10
MAYBBTB03715.1EA 0 EUR
MAYBBTB03715.1 MAYBBTB03715.10
3-Chlor-4-fluorbenzoesäure
3-Chloro-4-fluorobenzoic acid
Formel: C₇H₄ClFO₂
Molecular Weight: 174.56 g/mol
Siedepunkt: 273
Schmelzpunkt: 136
MDL: MFCD00042477
CAS-Nummer: 403-16-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR