5'-Fluor-2'-hydroxyacetophenon

Lieferant: Thermo Scientific

Synonyme: 1-(5-Fluoro-2-hydroxyphenyl)ethanone

BTB04130.1 BTB04130EA BTB04130EE
MAYBBTB04130.1EA 0 EUR
MAYBBTB04130.1 MAYBBTB04130EA MAYBBTB04130EE
5'-Fluor-2'-hydroxyacetophenon
5'-Fluor-2'-hydroxyacetophenon
Formel: C₈H₇FO₂
Molecular Weight: 154.14 g/mol
Siedepunkt: 182
Schmelzpunkt: 56
MDL: MFCD00011668
CAS-Nummer: 394-32-1
EINECS: 206-893-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR