2,6-Dibrom-4-cyclohexylanilin

Lieferant: Thermo Scientific

BTB04641.50
MAYBBTB04641.50EA 0 EUR
MAYBBTB04641.50
2,6-Dibrom-4-cyclohexylanilin
2,6-Dibrom-4-cyclohexylanilin
Formel: C₁₂H₁₅Br₂N
Molecular Weight: 333,07 g/mol
CAS-Nummer: 175135-11-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR