4-Brom-2,6-xylidin
Lieferant: Thermo Scientific
Synonyme:
4-Brom-2,6-dimethylanilin
BTB04886.1
BTB04886.10
BTB04886.50
MAYBBTB04886.1EA
0
EUR
MAYBBTB04886.1
MAYBBTB04886.10
MAYBBTB04886.50
4-Brom-2,6-xylidin
4-Brom-2,6-xylidin
Formel:
C₈H₁₀BrN Molecular Weight: 200.08 g/mol Siedepunkt: 120 °C (0.2 mmHg) Schmelzpunkt: 50 |
MDL:
MFCD00007826 CAS-Nummer: 24596-19-8 EINECS: 246-337-9 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...