4-Brom-2,6-xylidin

Lieferant: Thermo Scientific

Synonyme: 4-Brom-2,6-dimethylanilin

BTB04886.1 BTB04886.10 BTB04886.50
MAYBBTB04886.1EA 0 EUR
MAYBBTB04886.1 MAYBBTB04886.10 MAYBBTB04886.50
4-Brom-2,6-xylidin
4-Brom-2,6-xylidin
Formel: C₈H₁₀BrN
Molecular Weight: 200.08 g/mol
Siedepunkt: 120 °C (0.2 mmHg)
Schmelzpunkt: 50
MDL: MFCD00007826
CAS-Nummer: 24596-19-8
EINECS: 246-337-9
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR