2-[(2,6-Dichlorbenzyl)oxy]-5-nitrobenzaldehyd

Lieferant: Thermo Scientific

BTB05370.1
MAYBBTB05370.1EA 0 EUR
MAYBBTB05370.1
2-[(2,6-Dichlorbenzyl)oxy]-5-nitrobenzaldehyd
2-[(2,6-Dichlorbenzyl)oxy]-5-nitrobenzaldehyd
Formel: C₁₄H₉Cl₂NO₄
CAS-Nummer: 219539-46-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR