1,2-Dichlor-4-iodbenzol

Lieferant: Thermo Scientific

Synonyme: 3,4-Dichloroiodobenzene

BTB06155.1 BTB06155EA BTB06155.50
MAYBBTB06155.1EA 0 EUR
MAYBBTB06155.1 MAYBBTB06155EA MAYBBTB06155.50
1,2-Dichlor-4-iodbenzol
1,2-Dichlor-4-iodbenzol
Formel: C₆H₃Cl₂I
MDL: MFCD00019014
CAS-Nummer: 20555-91-3
EINECS: 243-874-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR