ω-Brom-3,4-difluoracetophenon

Lieferant: Thermo Scientific

Synonyme: 2-Brom-3',4'-difluoracetophenon, α-Brom-3,4-difluoracetophenon, 3,4-Difluorphenacylbromid, 2-Brom-1-(3,4-difluorphenyl)ethan-1-on

BTB06440.0005
MAYBBTB06440.0005EA 0 EUR
MAYBBTB06440.0005
ω-Brom-3,4-difluoracetophenon
ω-Brom-3,4-difluoracetophenon
Formel: C₈H₅BrF₂O
Molecular Weight: 235,03 g/mol
MDL: MFCD00085010
CAS-Nummer: 40706-98-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR