N-(2,4-Dimethoxyphenyl)-2-chloracetamid

Lieferant: Thermo Scientific

Synonyme: N-(Chloroacetyl)-2,4-dimethoxyaniline

BTB06598.1
MAYBBTB06598.1EA 0 EUR
MAYBBTB06598.1
N-(2,4-Dimethoxyphenyl)-2-chloracetamid
N-(2,4-Dimethoxyphenyl)-2-chloracetamid
Formel: C₁₀H₁₂ClNO₃
CAS-Nummer: 101908-41-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR