3,4-Dimethyl-o-phenylendiamin
Lieferant: Thermo Scientific
Synonyme:
1,2-Diamino-3,4-dimethylbenzol, 3,4-Dimethylbenzol-1,2-diamin, 3,4-Dimethyl-1,2-phenylendiamin
BTB06728.1
BTB06728.10
BTB06728.50
MAYBBTB06728.1EA
0
EUR
MAYBBTB06728.1
MAYBBTB06728.10
MAYBBTB06728.50
3,4-Dimethyl-o-phenylendiamin
3,4-Dimethyl-o-phenylendiamin
Formel:
C₈H₁₂N₂ Siedepunkt: 287 °C (1013 hPa) Schmelzpunkt: 89 °C |
MDL:
MFCD00051709 CAS-Nummer: 41927-01-9 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...