4-Amino-3-chlor-5-nitrobenzotrifluorid

Lieferant: Thermo Scientific

Synonyme: 2-Chloro-6-nitro-4-(trifluoromethyl)aniline

BTB06986.50
MAYBBTB06986.50EA 0 EUR
MAYBBTB06986.50
4-Amino-3-chlor-5-nitrobenzotrifluorid
4-Amino-3-chlor-5-nitrobenzotrifluorid
Formel: C₇H₄ClF₃N₂O₂
MDL: MFCD00042153
CAS-Nummer: 57729-79-0
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR