6-Oxo-1,4,5,6-tetrahydropyridazin-3-carbonsäure

Lieferant: Thermo Scientific

BTB07054DA BTB07054EA BTB07054EE
MAYBBTB07054DAEA 0 EUR
MAYBBTB07054DA MAYBBTB07054EA MAYBBTB07054EE
6-Oxo-1,4,5,6-tetrahydropyridazin-3-carbonsäure
6-Oxo-1,4,5,6-tetrahydropyridazin-3-carbonsäure
Formel: C₅H₆N₂O₃
Molecular Weight: 142.12 g/mol
MDL: MFCD00052030
CAS-Nummer: 27372-38-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR