4,5-Dichlor-o-phenylendiamin
Lieferant: Thermo Scientific
Synonyme:
1,2-Diamino-4,5-dichlorbenzol, 4,5-Dichlor-1,2-phenylendiamin, 4,5-Dichlorbenzol-1,2-diamin
BTB07389.1
BTB07389.10
BTB07389.50
MAYBBTB07389.1EA
0
EUR
MAYBBTB07389.1
MAYBBTB07389.10
MAYBBTB07389.50
4,5-Dichlor-o-phenylendiamin
4,5-Dichlor-o-phenylendiamin
Formel:
C₆H₆Cl₂N₂ Siedepunkt: 305 °C (1013 hPa) Schmelzpunkt: 163 °C |
MDL:
MFCD00007723 CAS-Nummer: 5348-42-5 EINECS: 226-305-0 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...