3-Chlor-2-benzo[b]thiophencarbonsäureamid

Lieferant: Thermo Scientific

BTB07540.50
MAYBBTB07540.50EA 0 EUR
MAYBBTB07540.50
3-Chlor-2-benzo[b]thiophencarbonsäureamid
3-Chlor-2-benzo[b]thiophencarbonsäureamid
Formel: C₉H₆ClNOS
Molecular Weight: 211,67 g/mol
MDL: MFCD00067793
CAS-Nummer: 21211-09-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR