2-(4-Fluorphenoxy)-3-nitropyridin

Lieferant: Thermo Scientific

BTB08014.1
MAYBBTB08014.1EA 0 EUR
MAYBBTB08014.1
2-(4-Fluorphenoxy)-3-nitropyridin
2-(4-Fluorophenoxy)-3-nitropyridine
Formel: C₁₁H₇FN₂O₃
MDL: MFCD00067801
CAS-Nummer: 147143-58-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR