2-(Phenylthio)nicotinsäure
Lieferant: Thermo Scientific
BTB08722DA
BTB08722EA
MAYBBTB08722DAEA
0
EUR
MAYBBTB08722DA
MAYBBTB08722EA
2-(Phenylthio)nicotinsäure
2-(Phenylthio)nicotinsäure
Formel:
C₁₂H₉NO₂S Molecular Weight: 231.2704 g/mol |
MDL:
MFCD00052110 CAS-Nummer: 35620-72-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...