2-(Phenylthio)nicotinsäure

Lieferant: Thermo Scientific

BTB08722DA BTB08722EA
MAYBBTB08722DAEA 0 EUR
MAYBBTB08722DA MAYBBTB08722EA
2-(Phenylthio)nicotinsäure
2-(Phenylthio)nicotinsäure
Formel: C₁₂H₉NO₂S
Molecular Weight: 231.2704 g/mol
MDL: MFCD00052110
CAS-Nummer: 35620-72-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR