2-(4-Methylphenylthio)nicotinsäure

Lieferant: Thermo Scientific

BTB08723.50
MAYBBTB08723.50EA 0 EUR
MAYBBTB08723.50
2-(4-Methylphenylthio)nicotinsäure
2-(4-Methylphenylthio)nicotinsäure
Formel: C₁₃H₁₁NO₂S
MDL: MFCD00052111
CAS-Nummer: 955-53-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR