2-Chlor-6-(trifluormethyl)pyridin

Lieferant: Thermo Scientific

BTB09171.250 BTB09171.1
MAYBBTB09171.250EA 0 EUR
MAYBBTB09171.250 MAYBBTB09171.1
2-Chlor-6-(trifluormethyl)pyridin
2-Chloro-6-(trifluoromethyl)pyridine
Formel: C₆H₃ClF₃N
MDL: MFCD00042226
CAS-Nummer: 39890-95-4
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR