Methyl-4-amino-2-methoxybenzoat

Lieferant: Thermo Scientific

Synonyme: 4-Amino-2-methoxybenzoesäuremethylester

BTB09531.1
MAYBBTB09531.1EA 0 EUR
MAYBBTB09531.1
Methyl-4-amino-2-methoxybenzoat
Methyl-4-amino-2-methoxybenzoat
Formel: C₉H₁₁NO₃
Molecular Weight: 181.19 g/mol
MDL: MFCD00017202
CAS-Nummer: 27492-84-8
EINECS: 248-494-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR