2-(4-Chlor-2-methylphenoxy)-3-nitropyridin

Lieferant: Thermo Scientific

BTB09540.50
MAYBBTB09540.50EA 0 EUR
MAYBBTB09540.50
2-(4-Chlor-2-methylphenoxy)-3-nitropyridin
2-(4-Chloro-2-methylphenoxy)-3-nitropyridine
Formel: C₁₂H₉ClN₂O₃
MDL: MFCD00096760
CAS-Nummer: 465514-31-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR