1-(2-Methoxy-4-nitrophenyl)pyrrolidin

Lieferant: Thermo Scientific

BTB09574.250 BTB09574DA
MAYBBTB09574.250EA 0 EUR
MAYBBTB09574.250 MAYBBTB09574DA
1-(2-Methoxy-4-nitrophenyl)pyrrolidin
1-(2-Methoxy-4-nitrophenyl)pyrrolidin
Formel: C₁₁H₁₄N₂O₃
MDL: MFCD00096778
CAS-Nummer: 67828-57-3
EINECS: 267-278-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR