1-Brom-4-pentylbenzol
Lieferant: Thermo Scientific
Synonyme:
4-Brom-pentylbenzol, (4-Bromphenyl)pentan, p-Brom-pentylbenzol
BTB09867.10
BTB09867.50
MAYBBTB09867.10EA
0
EUR
MAYBBTB09867.10
MAYBBTB09867.50
1-Brom-4-pentylbenzol
1-Brom-4-pentylbenzol
Formel:
C₁₁H₁₅Br Molecular Weight: 227.14 g/mol |
MDL:
MFCD00061113 CAS-Nummer: 51554-95-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...